Evaluate analysis results
You can evaluate analysis results in Ion Reporter™ Software by comparing multiple analyses. For example, you might compare new analysis results to known accurate analysis results.
Note: Comparison and visualization are not supported for Metagenomics analyses, or for analyses with a status of Failed or In-Progress. Tumor-normal analyses, which has only one set of variants for the tumor sample, and does not report variants for the normal sample, also cannot be visualized.
Visualization of a single trio or paired analysis is supported, because trio and paired analyses each contain more than one set of variants.
You can review the results in various ways. For more information, see Compare results of single or multiple analyses.