JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Allele View Allele View is an allele-centric view that displays each allele as a single row in the Ion Reporter™ Software and in an individual line in the VCF file output. Related Topics Parent topic: Locus View versus Allele View of variants ×