Show variants for a single chromosome
You can refine analysis results to show only those variants that are called on a single chromosome in Ion Reporter Software.
  1. In the Analyses tab, click Overview.
  2. In the Analyses screen, select the row of your analysis of interest.
  3. In the Analysis Results screen, in the Filter Options pane, navigate to the Chromosome list, then select the chromosome of interest.
The number of Filtered In Variants reflects only the variants on the selected chromosome. Because variants called on other chromosomes are considered to be "not displayed" instead of "filtered out", their variant totals are not added to the Filtered Out Variants for the analysis results.