You can add a classification to one or
more of the variants that are in analysis results in Ion Reporter™ Software. Classifications can be added to
individual variants, or to a list of selected variants. When a variant is classified,
the classification for the variant can be viewed in analysis results that include the
variant. Variant classifications are also added to final reports that are published from
the analysis.
Click the
Analyses tab
to open the list of analyses, then click an analysis of interest.
The Analysis Results table opens.
Classify variants individually, or apply the same classification to multiple
variants.
To classify variants individually, in the Classification column, use the
dropdown menu to select one of the following classifications for each
variant of interest: Unclassified, Unknown, Benign, Suspected Benign, Likely
Benign, Deleterious, Suspected Deleterious, Pathogenic, Likely Pathogenic,
VUS, Uncertain Significance, Technical Artifact.
To classify multiple variants with the
same classification, select the variants to which you want to add the
classification, then click Selected variants
> Classify variants. Then, select a classification from the dropdown list and
click
Confirm.