View original source of annotation information
You can view the original source of annotation information in Ion Reporter™ Software.
  1. In the Analyses tab, click Overview.
  2. Click the name of the analysis to open the Analysis Results table.
  3. In the Genes column, click the link. The link opens to the HUGO Gene Nomenclature Committee (HGNC) website.