The Analysis Results table
includes details that describe the results of the analysis, samples, analysis workflows,
and software versions that were used to create the analyses in Ion Reporter™ Software.
When you open an analysis, the information that is shown in the
Analysis Results table
depends on which workflow and annotation set is applied when the analysis is launched.
You can customize the table to reflect the information that is most relevant to your
research. To do so, create a table preference, or drag and drop the columns to change
the order in which they appear.