This filter can be added to filter chains in the software to return copy number variants (CNV) with confidence levels between 10 and 10,000,000 and other variants.
Filter conditions include the ability to set the CNV confidence range from 10 to 1.0E7. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter returns copy number variants with high confidence levels and other variants.