Lock a sample
You can lock a sample to prevent any user from editing the sample in Ion Reporter™ Software.
  1. In the Samples tab, in the Overview screen, select the sample in the table, then click Actions Lock.
  2. In the Lock Sample dialog box, click Yes to confirm that you want to lock the sample.
The locked sample is designated with a  (Lock) in the Samples table.