Delete a sample
You can delete one or more samples in Ion Reporter™ Software. For example, you can remove low‑quality samples or remove samples that were transferred to other organizations or archived. Storage space can increase when you delete samples.
IMPORTANT! When you delete a sample, this action also deletes the data files that are associated with the sample. If the BAM or VCF file that contains the data are associated with more than one sample, you cannot delete the sample.
×