Quality Control (QC) metrics
Quality Control (QC) metrics can be viewed in QC reports and in final reports. The metrics are based on data that are included in an analysis. For example, metrics that describe fusions are available only for analyses that include fusions data. See Create a final report template from analysis results and Create a final report for more information how to add QC metric to final reports. See Quality Control (QC) reports more information about QC reports.
The following fields are included in each of the *_coverage_statistics.txt files.
Column |
Description |
---|---|
The amplicon ID or gene name (if any). |
|
Usually the chromosome that contains the amplicon, gene, or hotspot region. |
|
The initial position of the amplicon, gene, or hotspot region. |
|
The end position of the amplicon, gene, or hotspot region. |
|
The number of reads with zero coverage. |
|
The number of reads with low coverage. |
|
The number of reads with medium coverage. |
|
The number of reads with high coverage. |
|
The region size (in bases) of the amplicon, gene, or hotspot region. |
|
The minimum coverage of any base in the amplicon, gene, or hotspot region. |
|
The maximum coverage in the amplicon, gene, or hotspot region. |
|
The average coverage in the amplicon, gene, or hotspot region. |
|
The number of bases with at least 1X coverage in the amplicon, gene, or hotspot region. |
|
The average base quality value. |