Ion Reporter™ Software generates Venn diagrams to display the
number of variants that are unique and common between analyses. You can use Venn diagrams to
limit visualizations to subsets of variants. For more information, see
Samples Venn diagram in Filter Options.
Venn diagrams are
generated for up to three similar analyses. Venn diagram generation in the software is
analysis workflow, research application, and sample group dependent.
Venn diagrams are
generated with following single analysis visualizations:
-
One DNA
Paired analysis.
-
One DNA
Trio analysis.
Venn diagrams are generated with multianalysis visualizations of the following
two analyses:
-
One DNA
Single analysis and
one DNA
Tumor–Normal
analysis.
-
One DNA
Single analysis and
one DNA and Fusions
analysis.
-
One DNA
Single analysis and
one DNA
Paired analysis.
-
One DNA and Fusions
analysis and one
Fusions
analysis.
-
One DNA
Tumor–Normal
analysis and one DNA
Paired analysis.
-
Two DNA
Single analyses.
-
Two DNA and Fusions
analyses.
-
Two DNA
Tumor–Normal
analyses.
-
Two Fusions
analyses.
-
Two
Annotate Variants
analyses.
Venn diagrams are generated with multianalysis visualizations of the following
three analyses:
-
One DNA
Single analysis and
two DNA
Tumor–Normal
analyses.
-
One DNA
Tumor–Normal
analysis and two DNA
Single analyses.
-
Three DNA
Single analyses.
-
Three
DNA and Fusions
analyses.
-
Three DNA
Tumor–Normal
analyses.
-
Three Fusions
analyses.
-
Three
Annotate Variants
analyses.
Venn diagrams are not generated for Ion AmpliSeq™ HD, Tagseq, immune repertoire, metagenomics,
and microbiome analysis workflows,