Samples Venn diagram in Filter Options
Use the Samples Venn diagram to limit the visualization of the selected analyses to a subset of variants. The numbers in the overlapping areas of the diagram represent variants that are common in the corresponding samples. The numbers in the non-overlapping areas represent the variants that are unique to that sample.
-
Click the numbers in the Venn diagram to select the variants for inclusion on the heat map.
-
Click Filter to update the heat map view to include only the selected variants. Click Clear to deselect all variants and restore the heat map to include all variants.
-
Sample and Analysis legend. The analysis name is listed in the parentheses.
-
Hover over a sample name to see the full sample name and the analysis to which the sample belongs. The analysis name is listed in the parentheses.