Chromosome filter in the Filter Options
In the Filter Options pane, you can use the Chromosome dropdown list to limit the visualization of the data to one chromosome. From the Chromosome dropdown list, select a chromosome number to visualize only the variants that are mapped to that chromosome. Select All to visualize all variants.