IRGV & Generate Report tab
You can view visualized analysis results in the IRGV & Generate Report tab.
-
Analysis Whole Genome View—Provides a genome-wide perspective of the data.
-
Selected Chromosome bar—To see the variant location on the selected chromosome, click the Whole Genome View image.
-
Proband Coverage Ratio—Type in the chromosome coordinates, or click the Karyo view to see discrete data for Copy Number calls.
-
Reference—Lists the genomic reference that is used for the analysis.
-
Shows annotation source results. To rearrange the annotation source tracks, click
(Settings).
-
Chart View Options—Toggle between a DNA View or RNA View. Select RNA View for fusion analyses to get a menu to search for individual fusions in the panel.
-
Sample/Analysis Summary—Sort by Sample number and rearrange the analysis rows on the left.
-
Search—Enter chromosome coordinates or gene names to find data.
-
Filter chain—Apply a filter chain and immediately view its results. You cannot save the results of the applied filter chain to the visualization. You can also create a new filter chain. For more information, see Filters and filter chains.
-
MAPD filter—An analysis sample-wide metric of noise. You can adjust it to determine when a particular sample has met the threshold that was set for the MAPD filter.
-
IGV/Export—Use multiple export options, including an export to IGV, and access to preferences. For more information, see Visualize variants with IGV and Set IRGV preferences.