JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Query by sample name API call You can query by sample name using this API call: /api/v1/analysis?format=json&name=[samplename]&type=sample This call returns a list of analysis names performed on the sample, ordered by start date. Related Topics Parent topic: Ion Reporter™ Software web services API ×