Compare the immune repertoire between samples
Ion Reporter™ Software can perform multisample (or cross‑sample) analysis to compare the immune repertoire between samples.
  1. Under the Analyses tab, in the Overview screen, select the appropriate analysis workflow from the Workflow filter.
  2. Select the adjacent checkboxes to select 2 or more results files from the available list of analyses, then click Visualize to generate a clone summary table.
  3. Select samples for a multisample analysis to generate comparison plots.
    Option
    Description
    Select 2 samples, then click Compare Samples.
    An interactive scatter plot for the selected samples is shown in the Comparison Plots tab.
    Select 2 or more samples, then click Compare Samples.
    A Jaccard Similarity Index for the selected samples is shown in the Comparison Plots tab.