Download clone summary and lineage summary files
-
To download the Clone Summary metrics file.
-
In the Immune Repertoire Results screen, Sample Results tab, select Clone Summary from the Views list.
-
Click Download Clone Summary.
The clone summary CSV file downloads automatically.
-
Open the .clone_summary CSV file to view the additional information that is included in the spreadsheet.
-
To download the Lineage Summary metrics file.
-
In the Immune Repertoire Results screen, Sample Results tab, select Lineage Summary from the Views list.
-
Click Download Lineage Summary.
The lineage summary CSV file downloads automatically.
-
Open the .lineage_summary CSV file to view the additional information that is included in the spreadsheet.
×