Final reports
You can create final reports directly from analysis results in Ion Reporter™ Software. You can customize the reports to include information about variants of your choice, and report sections that you can reorder, add and remove dynamically. Each section contains details about an analysis, so you can include information to meet requirement needs.
For example, reports can record information about the analysis workflow used, the annotations and filter chains that were applied to the analysis, the report generation date, and the name of the person who launched the analysis.
The reports can include sections for the samples used in the analysis, the genomic variants, and annotations for each variant. For example, the reports reflect the notes and classifications, quality control metrics, and background information for the analysis. Other sections that you can add are disclaimers, comments, electronic signatures, and images.
You can use system templates or create custom templates for these reports, and add a report template to an Ion Reporter™ Software analysis workflow to reduce report setup time.
You can create a final report in different ways:
-
Design and create a final report directly from a completed analysis. Customize the content that you include in the report and its organization, and make changes dynamically in a report preview. For more information, see Create a final report.
-
Predesign a final report template that you can attach to an analysis workflow. For more information, see Create a final report template from analysis results.
-
Predesign a final report template as an Ion Reporter™ Software workflow preset. The resulting report is not as customizable as a final report template that is created from an open analysis. For more information, see Create a final report template workflow preset.