You can delete unwanted custom final report templates.
In the Workflows tab, click Presets.
Select Final Report Templates from the list.
Select the row of the final report template that you want to delete, then click
(Actions) > Delete.
The final report template is now removed from the list of available templates in the Workflow Presets screen and can no longer be added to an Ion Reporter™ Software analysis workflow.