You can edit custom final report templates that exist in the user account.
In the Workflows tab, click Presets.
Select Final Report Templates from the list.
Select the row of the final report template that you want to edit, then click (Actions) > Edit.
You can change the name and the description, and select sections to include or exclude, then drag and drop Reported Variants details.
In the Footer Section, you can remove the Thermo Fisher Scientific logo from the report footer and add a custom logo. Acceptable formats are PNG, GIF, and JPG.
Click Remove to remove the Thermo Fisher Scientific logo from the report footer.
Click Upload Logo, then click Select File to browse to a custom logo file.
Select the file, then click Open to include the logo in the report footer.
Click Save.
If the template is included in an Ion Reporter™ Software analysis workflow, it is now available for use when you review analysis results. If it was not, you must add the template to an analysis workflow. For more information, see Add a report template to an analysis workflow.