V-gene usage and largest clone frequency plot example
IGH V-gene usage and largest clone frequency
Select Largest Clone Frequency for a view of a spectratyping plot that highlights the frequency of the largest clone for each variable gene-CDR3 nucleotide length combination.
This is an example of an IGH V-gene usage and largest clone frequency spectratyping plot from an Oncomine™ BCR IGH‑LR Assay analysis. Circle color indicates the frequency of the largest clone having a particular variable gene-CDR3 nucleotide length combination. Dark color indicates the presence of expanded clones. This is a polyclonal repertoire that lacks highly expanded clones. You can further partition the clones by isotype in Ion Reporter™ Software, with the buttons that are above the interactive spectratyping plot.