Immune repertoire analysis results report Variable gene-Joining gene (VJ-gene) combinations for
identified clones. Select
VJ‑ gene usage heatmap
from the Views
list to see heatmaps that represent the results.
VJ‑gene usage heatmap example
Heatmap illustrating the frequency of particular Variable gene-Joining gene combinations for identified clones. Frequencies are log transformed with pseudocount added. Over-represented Variable-Joining gene combinations (white) can indicate the presence of a highly expanded clone.