|
Import
|
The Import User is the person in the Ion Reporter™ Software organization who can import and define samples. Specific access includes:
|
|
Analyze
|
The Analyze User is the person in the Ion Reporter™ Software organization who can create analysis workflows and launch analyses. Specific access includes:
-
View samples.
-
Define samples.
-
Create or modify analysis workflows.
-
View analysis workflows.
-
Launch an analysis.
-
View analyses list.
-
Review and filter analysis results.
-
Access the Samples, Analyses, and Workflows tabs.
|
|
Report
|
The Report User is the person in the Ion Reporter™ Software organization who can generate reports. Specific access includes:
-
View the analyses list (and view and classify results).
-
Generate reports and sign off on final and visualization reports using an electronic signature.
-
Select and classify variants.
-
Access the Analyses tab.
|
|
Admin
|
The Admin is the person in the Ion Reporter™ Software organization who can manage user access. The Admin user can:
-
View samples.
-
View analysis workflows.
-
View the analyses list (but not view results).
-
Access the Samples, Analyses, Workflows, and Admin tabs.
-
Create and modify users.
-
Download and install plugins.
-
Update software on Ion Reporter™ Server. For more information, see Ion Reporter™ Server Upgrade Instructions User Bulletin (Pub. No. MAN0017841).
|