Create an organization on Ion Reporter™ Server
The first user on Ion Reporter™ Server can create an organization. An organization is more specific than an institute name. It defines a group of users who work together and share samples, data, analyses, and results.
When you create an organization, you automatically are the first administrator-level user of that organization. You can then begin to enter or import data into the software and invite other Ion Reporter™ Server users to the organization to share data and results.
You can belong to only one organization at a time. If you are already a member of an organization on Ion Reporter™ Server, you will not be able to create a new organization until you are removed as a user from your current organization.
- On the software sign-in screen click, click Register new account.
-
In the Request Account dialog box, enter the following information: organization name, organization address, your first and last name, email, phone number.
If the Ion Reporter™ Server user is connected to the Internet, you must select the checkbox in the dialog box.
-
Click Submit.
A new organization is created and you become the organization administrator. You will receive an email with a link to set the password.
- Click the link in the email and set the password.
- After the password is set, you can sign in using the new password. The first time you sign in, you must accept the Terms and Conditions.