This image shows an example of the
Analysis Results table
for a run in which no fusions are called.
Note: When you sort on the
Gene column,
the software sorts on both of the genes involved in the fusion. As a result, the
lexicographically smaller gene is used for an ascending sort, while the
lexicographically larger gene is used for a descending sort.