Amplicon coverage report metrics
The amplicon coverage report includes the following metrics:
Metric |
Description |
---|---|
The amplicon name as defined in the BED file for the amplicon region. |
|
Chromosome |
|
The start coordinate of the amplicon. |
|
the end coordinate of the amplicon. |
|
The attribute in the BED file for the amplicon. |
|
The number of assigned reads from end to end for the amplicon region. |
|
The number of assigned forward strand reads from end to end for the amplicon region. |
|
The number of assigned reverse strand reads from end to end for the amplicon region. |
|
The total number of reads assigned to this amplicon. This value is equal to Fwd + Rev. |
|
The number of forward strand reads assigned to this amplicon. This value may differ from the reads for end to end. |
|
The number of reverse strand reads assigned to this amplicon. This value may differ from the reads for end to end. |