For multi-pool RNA pools, you can specify the minimum number of pools in a Sample that are required to pass QC to qualify the Sample as Valid, then proceed with the Analysis. If the number of valid pools in any sample is less than the specified threshold, that Sample does not pass QC. Valid pools are determined by the total valid mapped reads per pool.
Example: If a panel has two pools, use value=2 to specify that both pools need to have a sufficient number of reads in order to qualify that sample. Similarly use value=1 to proceed with the analysis even if one of the pools failed.