Pfam filter
This filter can be added to filter chains in the software to detect protein domain families in the coded proteins as defined by the Pfam database. The specific filters are as follows.
Pfam (31)
Pfam (32)
Filter conditions include the ability to include all (In), none (Not In), or specific annotations, and to include or exclude unannotated variants in filtered analysis results.
The filter returns variants in the samples that match the variants in the Pfam database.