You can lock custom final report templates so that they cannot be altered.
In the Workflows tab, click Presets.
Select Final Report Templates from the list.
Select the row of the final report template that you want to lock, then click
(Actions) > Lock.
A lock icon appears the row of the final report template in the
Workflow Presets
screen. You can add this report template to an analysis workflow. For more
information, see Add a report template to an analysis workflow.