Predefined analysis workflows
Analysis workflows are sets of analysis components that you can use to automate the analysis of data. Predefined analysis workflows are optimized with parameters for use with specific products.
These predefined analysis workflows are organized into the following research categories:
Research category
Description
Exome
For use with Ion AmpliSeq™ Exome and TargetSeq Exome panels.
16S rRNA Profiling
For use with 16S Metagenomics panels and Ion AmpliSeq™ Microbiome Health Research panels.
Oncology–Liquid Biopsy
For use with a variety of Ion AmpliSeq™ and Ion AmpliSeq™ HD, and Oncomine™ cancer panels and assays.
Reproductive
For use with Ion ReproSeq™ and other PGS whole genome assays, and the Ion AmpliSeq™ Ion CarrierSeq™ ECS Panel.
Immunology
For use with a variety of Ion AmpliSeq™ cancer panels and Oncomine™ immunology assays.
For use with a variety of Ion AmpliSeq™ and Ion AmpliSeq™ HD, and Oncomine™ cancer panels and assays.
Oncology–ImmunoOncology
For use with a variety of Ion AmpliSeq™ and Oncomine™ cancer panels and assays.
Annotate Variants
For use with VCF samples as the input to annotate variants.
Oncology–HemeOnc
For use with a variety of Ion AmpliSeq™ and Ion AmpliSeq™ HD, and Oncomine™ cancer panels and assays.
Oncology–Solid Tumor
For use with a variety of Ion AmpliSeq™ and Ion AmpliSeq™ HD, and Oncomine™ cancer panels and assays.
Carrier Screening
For use with the Ion AmpliSeq™ Ion CarrierSeq™ ECS Panel.
Inherited Disease
For use with a variety of Ion AmpliSeq™ and Oncomine™ cancer assays.
Plugin
Inserts plugin code into a workflow and allows a plugin to be included as the last step or the only step in when an analysis workflow is launched.