Visualize RNA exon variants
You can view intragenic exon rearrangements, such as exon deletions, exon skipping events, and alternate splice transcripts graphically in Ion Reporter™ Software. You can view RNA Exon Variant graphs from a single analysis or from multiple analyses simultaneously. If multiple analyses are compared, the data for each sample is summarized in a separate bar graph. For more information, see Compare results of single or multiple analyses. RNA Exon Variant graphs are available for all analyses that use RNA or Fusion analysis workflows.
RNA Exon Variant graphs show variant read counts, normalized to the wild type, for specific exon variants.