Time series visualizations
You can track and visualize variants over time in Ion Reporter™ Software. With time series visualizations, you can view trends and changes for variant alleles over time. For example, you could graph the percent frequency of TP53 c2369C>T at regular intervals from specimens collected from a single individual. Time series graphs show both qualitative (such as no call instances) and quantitative (such as the allele ratio) results for variants. You can remove and restore variants dynamically in the graphs that are shown in the software, and adjust how data are organized in the graph before you generate the chart. Customization of the graphs allows you to visualize the most relevant information.
To visualize results with a time series graph, you must select analyses that use the same analyses workflows. The name of the analysis workflow, the version number, and the revision number, if applicable, must be identical.
Time series graphs are supported for the following variant types:
|
Type of graph |
Supported variants |
|---|---|
|
Fusion |
|
|
SNVs/Indel |
|
|
CNV |
|