Minimum Mean Read length
Mean Read Length is a QC metric available for the gene fusion analyses that indicates whether the mean read length computed from all the reads in the sample is less than the specified threshold. If the mean read length is below the threshold, the sample does not pass the quality control.
A 50bp minimum mean read length is recommended to avoid the possibility of missing real fusions (false negatives) and reporting false positive fusions.