Download filtered variants
- In the Analyses tab, click Overview, then select the analysis or analyses that contains the filtered variants.
For a single analysis, a compressed directory that is named analysis name_Filtered.zip is downloaded. For multiple analyses, a compressed directory that is named batch_analysis_results_date_system-generated-number_Filtered.zip is downloaded.
- To download the ZIP file from Ion Reporter™ Software:
- Extract the contents of the directory to access the folders and files.
Folder
Description of contents
Gene_deletions.xls, a file that contains information about whole gene deletions and amplicon coverage that are found in BRCA analyses. Values in the third column of the file indicate the following information for the BRCA1 and BRCA2 genes in the sample.
SQCFAIL—Calculated overall variation between amplicon coverage is too high.
LGD=<score>—Large Gene Deletion with Phred scale score = <score>.
WGD=<score>—Whole Gene Deletion with Phred scale score = <score> (max score = 100).
The Phred scale score is must >=40, for the WGD (subtype = GeneCNV) to be reported in the VCF file. Large gene deletions are not reported in the VCF file.
HRR_Results
Contains a .TSV file that contains a summary of the HRR results.
signature_prediction
The signature_prediction folder contains:
Torrent Suite™ Software Quality Control (QC files. For example, bam_file_basename>.ionstats_alignment.json, analysis.bfmask.stats, explog_final.txt, raw_peak_signal, InitLog.txt, basecaller_results/BaseCaller.json, basecaller_results/ datasets_basecaller.json, basecaller_results/ionstats_tf.json, basecaller_results/TFStats.json
RNAQCAndCalls.txt—contains information on the QC for fusion data.
TotalMappedFusionPanelReads.txt—provides number of total mapped fusion panel reads.
<analysis name>_QC.pdf—Report of QC metrics in PDF format. The same QC metrics are available in the Final Report of an analysis.
Contains VCF and TSV files for the CNV variants. You can open these files in Microsoft™ Excel™.
Note: If the Ion Reporter™ Software analysis includes notes, the notes are included in the files in a row named #analysisnotes.
Analysis_Settings —Contains a text file that describes settings used for the analysis. Open the file with a text editor.
Module_Configuration_Files—Contains configuration files for workflows, including VariantCallerActor.json, a JSON file that contains variant finding parameters used to customize variant calling in Ion Reporter™ Software and the settings used in the analysis. The file shows changes made to the parameters in the software. The file is present only for analyses that contain variants.
Files with data about microsatellite instability (MSI) markers. These markers can identify a form of genomic instability in the replication of repetitive DNA. MSI often occurs in tumor cells. It leads to the appearance of multiple alleles at microsatellite loci, which can be easily identified.
Summary.tsv—Provides a summary of results for MSI markers, as shown in the Analysis Results table, and additional information about the algorithm version that is used to calculate the Marker MSI Score and MSI Score.
Details.tsv—Provides marker-level information on microsatellite instability, such as individual marker MSI scores, and read coverage for individual MSI markers.
MSIQC.json —File contains "flags" and warning messages associated with MSI status that are used by customer support representatives.
If the Tumor Mutational Burden parameter is enabled, or an analysis workflow for the Tumor Mutation Load Assay is used, a Results folder is generated that contains:
filter_variants.tsv and somatic_variants.tsv—TSV files that contain post-filter and somatic variants.
<tmb report>.pdf—PDF report that contains tumor mutational load results.
PNG files that contain images of:
base_change_file.txt and somatic_mutation_substitution_context_summary.txt— four files that contain the data used to generate the substitution type and context of somatic mutations and substitution type and signature pattern of somatic mutations plots
deamination_metric.txt — contains the TMB score and other TMB metrics
Contains ZIP file of the plugin output directory.
Immune repertoire output
The CSV, PNG and PDF files that show the data for Immune repertoire analyses.
Files from a single metagenomics sample, or results from multiple samples that the software compares for Beta diversity, and files that contains reads for the analysis in FASTA format, and other TXT files for the analysis.
To download all results files for metagenomics analyses, use instead the Download results files for all samples link, shown in the Visualization/Downloads section of the metagenomics analysis results. For more information, see View interactive taxonomy charts for metagenomics analyses.