Custom analysis workflows
There are three ways to create custom analysis workflows in Ion Reporter™ Software.
-
Copy an existing analysis workflow, then edit its settings to create a custom analysis workflow. For more information, see Create a custom analysis workflow from an existing analysis workflow.
-
Using the workflow bar that walks you through the process, create a unique custom analysis workflow that contains unique settings. For more information, see Create a custom analysis workflow without predefined settings.
-
For Ion AmpliSeq™ HD panel files that are imported from AmpliSeq.com, copy the appropriate Ion AmpliSeq™ HD analysis workflow template, then use the template to make further changes to the custom analysis workflow, then save the analysis workflow before the custom workflow is used for analysis. For more information, see Create a custom analysis workflow for use with Ion AmpliSeq HD panels.