Workflow tab overview
You can access the list of available analysis workflows in the Workflows table, which is located in the Workflows tab, in the Overview screen.
  1. Workflows tab.
  2. The Overview screen in the Workflows tab. Click the screen name to toggle between the different screens.
    The Create screen. For more information, see Copy and edit an existing analysis workflow.
    The Presets screen. For more information, see Workflow presets.
  3. The Search box enables you to enter keywords or text strings to narrow the list of analysis workflows. For more information, see Search for analysis workflows.
  4. The Research Application filter enables you to search by application type.
  5. The Workflow list enables you to filter by analysis workflows that are predefined, user-defined, or tagged for IonReporterUploader plugin.
  6. The Target list enables you to filter by sequencing technology.
  7. Import Workflow enables you to upload a file to create an analysis workflow.
  8. Create Workflow enables you to create a new analysis workflow.
  9. The Research Category filter allows you search by application category.
  10. The Group list enables you to filter by sample grouping type.
  11. The Version filter enables you to search by Ion Reporter™ Software version.
  12. The Reference filter enables you to search by GRCh38 or hg19 references.
  13. Table columns with information about the analysis workflows.
  14. (Locked) Indicates that the analysis workflow is locked.
  15. Ion Factory Shipped If present, indicates that the analysis workflow is predefined with parameters for use with specific products.
  16. Details provides more information about the selected workflow.