JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Delete a Samples table preference You can delete a table preference that is no longer needed in Ion Reporter™ Software. In the Samples tab, click Overview. Click Preferences > Table Preferences. A menu of saved preferences appears. Select the preference that you want to delete from the menu. Click Preferences > Delete Table Preference. In the Confirm Delete, click Yes to delete the table preference. The table preference is deleted and the Samples table returns to the default order of columns. Related Topics Parent topic: Customize the Samples table ×