You can create Samples table preferences to make it easier to view the sample attributes and information. Table preferences allow you to include or exclude sample attributes, and drag then drop each column to the desired position in the table. When you create a Samples table preference, you can apply it at any time to the Samples table. You can also use Samples table preferences to toggle between different views of the Samples table.
In the Samples tab, click Overview.
Click Preferences > Select Columns.
In the Select Columns
dialog box, then select the sample attributes that you want to add to the
Samples table,
or deselect the sample attributes that you want to remove from the table.
Click Preferences > Save Table Preference As.
In the Save Table Preference dialog box, enter a name for the table preference, then click Save.