The hg19 and GRCh38 dbsnp versions in Ion Reporter™ Software 5.12 are both dbsnp versions 151. However, since the UCSC genome browser did not have the UCSC common genes available for dbSNP version 151 when Ion Reporter™ Software 5.12 was being released, UCSC common genes version 150 are used to set the UCSC common genes in Ion Reporter™ Software 5.12.