IsNewlyHomozygousRef
A proband variant is newly homozygous if it is homozygous but genotypes of both of the parents are heterozygous.
A proband variant is assigned the category IsNewlyHomozygousRef if all of the following are true:
- The variant is homozygous.
- The variant's allele is the same as the reference.
- The father's genotype is heterozygous at this variant position.
- The mother's genotype is heterozygous at this variant position.
- The frequency of the allele in the population is at most newly_homozygous_nonref_max_population_allele_frequency.
×