Change amino acid code
Ion Reporter™ Software gives you the option to annotate amino acid changes with the single-letter IUPAC amino acid codes, or the three-letter codes, based on your preference. By default, the three-letter code is used. To use the one-letter IUPAC code, you can change a parameter when you edit an analysis workflow. Then, for example, Val600Glu would be used in the annotation and displayed in the analysis results as V600E.
- In the Workflows tab, select the analysis workflow of interest, then click Edit.
- Click Parameters in the workflow bar.
-
In the Annotation section, under Use IUPAC Single Letter Code for Amino Acid, select the option that you want to use for amino acid changes:
Option
Description
True
Select to use the single-letter IUPAC code
False
Select to use the three-letter amino acid code
- Click Next.
- Review your selected options for the edited analysis workflow, then click Confirm.