Filter analyses
You can use filters on the Analyses table to narrow search results, or shorten the list of analyses.
  1. In the Analyses tab, click Overview.
  2. (Optional) You can add more columns to the table, then filter on those columns.
    1. Click PreferencesSelect Columns to add more columns.
    2. Add user-defined sample attributes or predefined analyses attributes as filters:
      • Click More Filters, then select one or more applicable filter categories from the list.
      • Click More Filters, type in a search string into the Find Filters field, then select one or more applicable filter categories.
      • Click More Filters, then click Select All to select all filter categories.
  3. Apply the filters:
    • Click the filter to expand the list, then select one or more specific filters. For example, select Chip Type to filter for sequencing data from an instrument that uses a specific type of sequencing chip.
    • Click the Version filter to view all analyses or select from the list to filter for specific versions.
    • Click the Workflow filter to view all analysis workflows or select from the list to filter for specific versions.
    • Click the filter to expand the list, then type search string in Find Filters. Select one or more specific filters. For example: select 530 to filter for sequencing data from an instrument that uses a Ion 560™ Chip.
    • Click the filter to expand the list, then click Select All to select all filters in a specific filter category.
    The contents of the Analyses table changes each time you select a filter or set of filters.
The filtered list of analyses is returned in the Analyses table.