JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Deamination filter This filter can be added to filter chains in the software to filter C:G>T:A SNVs that are detected at all allele frequencies. Filter conditions include the ability to exclude (Not In) or include (In) these variants in filtered analysis results. Related Topics Parent topic: Data type filters ×