Import a custom annotation source
You can use custom annotation source files in Ion Reporter™ Software. If you have annotation data that you want to use, you need to first create a new annotation set for the custom annotation source. Then, you can import the files into the new annotation set.
There are 4 types of annotation sources that you can import into Ion Reporter™ Software. The types of annotation sources and the file formats required for each annotation source are as follows.
Annotation source type
Description
File format for import
Genomic region
A set of regions
BED
Gene set
A set of genes
TXT
Preferred transcript set
A set of transcripts
TXT
VariantDB
A set of variants
VCF
  1. In the Workflows tab, click Presets.
  2. Click Create PresetAnnotation Set.
  3. Enter a descriptive Name for the custom annotation set.
  4. (Optional) Enter a description for the custom annotation set.
  5. Select a custom annotation type from the dropdown list:
    • Genomic Region (Custom)
    • Gene Set (Custom)
    • Preferred transcript set (Custom)
    • VariantDB (Custom)
  6. Click the Create New tab.
  7. Enter a descriptive Name and Version for the custom annotation source.
  8. (Optional) Enter a description for the custom annotation source.
  9. Click Select File, then find and select the custom annotation source file, then click Open.
  10. Click Save.
  11. The new custom annotation source is added to the custom annotation set.