The FusionSampleOverallCall allows the user to confirm whether a sample is a No Call, Positive, or Negative. The following logic is applied for the panels that have both targeted fusion isoforms and 5'/3' Imbalance Assays:
|
Fusion Isoforms
|
RET/ROS1/ALK Imbalance Score
|
Final Call
|
|
Positive
|
Positive
|
Positive
|
|
Positive
|
No Call/Negative
|
Positive
|
|
Negative
|
Positive
|
Positive
|
|
Negative
|
Positive
|
Positive
|
|
Negative
|
No Call
|
No Call
|
|
Negative
|
Negative
|
Negative
|