Fusion sample QC metrics
Fusion Sample QC, Fusion Overall Call, Total Mapped Fusion Panel Reads, Total Unmapped Reads, and Process Controls Total Reads are included in Ion Reporter™ Software.
These metrics are available in the Analysis Results screen for analyses that include fusions.
Note: In the Analysis Results screen, you can close the Filter Options on the right, to expand the table.