non-frameshift insertion or deletion
Insertion or deletion of the number of nucleotide base that are divisible by 3, hence, the inclusion or exclusion of amino acid in the protein translation from the original.