Get current results TSV file
getcurrentResultTsv
URL
api/v1/currentResultTsv?parameters
Description
Returns the Current Results TSV file path in the same column order as shown in the analysis results in Ion Reporter™ Software.
Method
GET
Request parameters
  • format.
    String. Optional.
    Response type, current version supports only json.
    format=json
  • type
    String, Optional.
    Valid values are sample, or analysis.
  • name
    String, Conditional.
    If type is sample, then name=NAME OF THE SAMPLE.
    If type is analysis, then name=NAME OF THE ANALYSIS.
If there are two analyses with the same name for two different versions of Ion Reporter™ Software, then the request returns two file paths for the TSV files. If the analysis is not found, the software returns an error with an explanation that the analysis was not found.
Example request
http://ionreporter.thermofisher.com/api/v1/currentResultTsv?format=json&name=xyzsampletest
Request headers
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
Returns
[
    {
        "report_published": "",
        "workflow": "Low-pass whole-genome aneuploidy w1.0",
        "data_links": {
            "current_result_tsv": "https://<server name>/api/v1/download?filePath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/Demo_Aneuploidy/Demo_Aneuploidy_20180419122112036/analysis_downloads/currentTsv/Demo_Aneuploidy_c1531_2018-04-19-12-26-20-867-2018-07-10-16-04-13-869.tsv"
        },
        "samples": {
            "PROBAND": "Demo Aneuploidy"
        },
        "variants_saved": "",
        "shared_with": [],
        "stage": "Send for Report Generation",
        "flagged": false,
        "name": "Demo Aneuploidy_c1531_2018-04-19-12-26-20-867",
        "id": "ff80818162dca67d0162dcabdd6b0001",
        "ion_reporter_version": "5.10",
        "started_by": "Ion User",
        "start_date": "April 19, 2018",
        "status": "SUCCESSFUL"
    }
]
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -v -k -X GET "https://server name/api/v1/currentResultTsv?format=json&name=BRCA_Plus_RR_jul29" -H "Content-Type:application/x-www-form-urlencoded" -H "A