Annotation sets created by users are displayed in the Annotation Source list. You can mix customer annotation sources with Ion Reporter™ Software predefined annotation sources to create your own custom annotation set. For more information, see Create annotation set workflow presets.
In the Workflows tab, click Presets.
Click Annotation Set > Annotation Source.
A table listing the custom annotation sources appears. You can use these custom annotation sources to create custom annotation sets.