This section provides details about the annotation scores used in Ion Reporter™ Software and how these scores are calculated.
SIFT versions
-
SIFT version—5.1.1
-
Protein database—UniprotTrEMBL, downloaded on July 11, 2011. Updated on March 14, 2014.
-
BLAST version—2.2.25. Updated to 2.2.26.
-
BLIMPS version—3.9